Learn More
Bachem pTH-Related Protein (1-34) (human, mouse, rat), 1mg, 112540-82-6, MFCD00081710 DFS Item
Manufacturer:  Bachem 4017147.1000
V.Paspaliaris et al. showed that daily administration of pTHrP (1-34) to rats was anabolic on bone by increasing bone formation. This treatment could inhibit the rapid decline in bone formation due to denervation.
Catalog No. 50-194-6982