Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSA V-TEC™ io1 Smart Hook Assembly Kit 36CSN

A self-retracting lifeline, specifically designed for lift work in warehouses and fulfillment centers

$971.00 - $1079.00

Specifications

Color Clear, Green
Diameter (English) 0.5 in.
Material Polycarbonate, SST
Product Type Self Retracting Lanyard
For Use With (Application) For lift work in warehouses and fulfillment centers
View More Specs

Products 2
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
Catalog Number Mfr. No. Length (English) Price Quantity & Availability  
17-200-200
View Documents
MSA
10213902
6 ft.
Each for $971.00
Add to cart
 
17-200-202
View Documents
MSA
10216248
10 ft.
Each for $1,079.00
Add to cart
 
Description

Description

  • Seat-belt alarm style system helps to improve safety compliance rates
    • Continues to sound until workers are connected in lift
  • ANSI
Specifications

Specifications

Clear, Green
Polycarbonate, SST
For lift work in warehouses and fulfillment centers
0.5 in.
Self Retracting Lanyard
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit