Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V-type proton ATPase subunit F Mouse anti-Human, DyLight 550, Clone: OTI1B8, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274863R
Description
V-type proton ATPase subunit F Monoclonal antibody specifically detects V-type proton ATPase subunit F in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
V-type proton ATPase subunit F | |
OTI1B8 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
ATP6V1F | |
Full length recombinant protein of human V-type proton ATPase subunit F (NP_004222) produced in E.coli. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ATP6S14adenosinetriphosphatase 14k chain, ATPase, H+ transporting, lysosomal 14kDa, V1 subunit F, H(+)-transporting two-sector ATPase, 14kD subunit, MGC117321, MGC126037, MGC126038, vacuolar ATP synthase subunit F, vacuolar proton pump F subunit, Vacuolar proton pump subunit F, VATFATPase, vacuolar, 14 kD, V-ATPase 14 kDa subunit, V-ATPase F subunit, V-ATPase subunit F, Vma7, V-type proton ATPase subunit F | |
Mouse | |
IgG2a | |
0.1 mL | |
Primary | |
9296.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title