Learn More
Apogee Designs LtdSupplier Diversity Partner V16 ELECTROPHORESIS SYS DFS Item
Manufacturer:  Apogee Designs LtdSupplier Diversity Partner 21070010
The Model V16 is designed to electrophorese a single 15 x 17cm vertical gel for the separation of proteins and nucleic acids. Features and components included: Features ; A neoprene gasket provides leak-free sealing of the glass plate assembly. ; Fits gels of different thicknesses with straight or notched lass plates. ; Binder clips for gel clamping are included. ; Safety lid includes attached power cords. ; S2 like integrated clamps hold the glass plate gel sandwich in place; no need for binder clips. An extensive list of precision machined Delrin (acetal) combs and spacers may be ordered separately allowing you to choose what's best for your needs.
Catalog No. 50-478-645