Learn More
Pyramex™ V2G-Plus™ Safety Eyewear
Click to view available options
Lens Tint:
Clear
Gray
Description
Specifications
Lens Type | H2X |
Certifications/Compliance | ANSI Z87.1,CSA Z94.3-07 |
Certifications/Compliance (Europe) | EN-166 |
Lens Tint | Gray |
Product Type | Goggle with Strap |
Frame Color | Black |
Weight (Metric) | 80 g |
Lens Material | Polycarbonate (front lens), Acetate (inner lens), PU foam |
Frame Material | TPU |