Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag, Armenian Hamster anti-Virus, Clone: SV5-P-K, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP2810360.2MG
Description
V5 Epitope Tag Monoclonal specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunoassay.Specifications
V5 Epitope Tag | |
Monoclonal | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunoassay | |
V5 epitope tag | |
Pk (V5) Epitope Tag (GKPIPNPLLGLDST). | |
0.2 mg | |
Primary | |
Virus | |
Purified |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen) | |
SV5-P-K | |
Unconjugated | |
PBS | |
Armenian Hamster | |
Protein A purified | |
RUO | |
Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction