Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag, Human anti-Virus, Clone: SV5-P-K, Novus Biologicals™

Human Monoclonal Antibody

$135.50 - $419.00

Specifications

Antigen V5 Epitope Tag
Clone SV5-P-K
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Host Species Human
Target Species Virus
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB014507
View Documents
Novus Biologicals
NBP2810350.025MG
0.025 mg
Each for $135.50
Add to cart
 
NB014508
View Documents
Novus Biologicals
NBP2810350.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Virus Samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Radioimmunoassay.
Specifications

Specifications

V5 Epitope Tag
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen)
Virus
Primary
SV5-P-K
Human
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit