Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag Mouse anti-Epitope Tag, Alexa Fluor 750, Clone: E10/V4RR, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP237825AF750
Description
V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunofluorescence.Specifications
V5 Epitope Tag | |
E10/V4RR | |
Western Blot, ELISA, Immunofluorescence | |
Purified, Conjugated | |
Mouse | |
IgG1 | |
0.1 mL | |
Primary | |
This product recognizes V5 (GKPIPNPLLGLDST) tagged fusion proteins. It has successfully been used in immunoprecipitation, ELISA, Western blotting, and immunofluorescence applications. |
Western Blot, ELISA, Immunofluorescence | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
V5 epitope tag | |
GKPIPNPLLGLDST (V5) synthetic peptide conjugated to KLH | |
Protein A purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Epitope Tag |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title