Learn More
V5 Epitope Tag Mouse anti-Virus, Alexa Fluor™ 350, Clone: SV5-P-K, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252703AF350
Catalog No. NP22703A30
Add to cart
Description
Description
V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Virus samples. It is validated for Western Blot,ELISA,Immunohistochemistry,Immunohistochemistry (Frozen),Immunofluorescence.
Specifications
Specifications
V5 Epitope Tag
SV5-P-K
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence
V5 epitope tag
Pk (V5) Epitope Tag (GKPIPNPLLGLDST).
0.1 mL
Primary
Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5.
ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Virus