Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag Antibody (SV5-Pk5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
$204.00 - $482.50
Specifications
Antigen | V5 Epitope Tag |
---|---|
Clone | SV5-Pk5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 |
Applications | Immunoprecipitation, Immunocytochemistry, ELISA, Immunofluorescence, Western Blot |
Description
V5 Epitope Tag Monoclonal specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
V5 Epitope Tag | |
1 mg/mL | |
Immunoprecipitation, Immunocytochemistry, ELISA, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Epitope Tags, V5 Epitope Tags | |
A recombinant peptide corresponding to the PK tag fused to a class II - restricted T helper cell epitope. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
SV5-Pk5 | |
Western Blot 1:100-1:500, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:2000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
V5 epitope tag | |
IgG1 | |
Protein A or G purified | |
Recognises a small epitope, termed Pk, present on the P/V proteins of the paramyxovirus, SV5. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title