Learn More
V5 Epitope Tag Rabbit anti-Many Species, Alexa Fluor 700, Clone: 1036H, R&D Systems™
Rabbit MonoclonalAntibody
Manufacturer:  R&D Systems FAB8926N100UG
Catalog No. FAB8926N100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
V5 Epitope Tag
Monoclonal
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
GKPIPNPLLGLDST Epitope Tag, Parainfluenza virus 5 V/P, Paramyxovirus SV5, V5 epitope tag
V5 epitope tag peptide Accession # Y138513
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Multi-Species
Flow Cytometry
1036H
Flow Cytometry 0.25-1 ug/10^6 cells
Y138513
Rabbit
IgG
RUO
Primary
Detects N-terminal and C-terminal V5 epitope-tagged proteins in Western blots.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)