Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
V5 Epitope Tag Antibody (SV5-P-K), Alexa Fluor™ 532 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252653AF532
Description
V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Epitope Tag samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence.Specifications
V5 Epitope Tag | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Rabbit | |
Protein A or G purified | |
Primary | |
Store at 4°C in the dark. | |
Epitope Tag |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence | |
SV5-P-K | |
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence | |
V5 epitope tag | |
Pk (V5) Epitope Tag (GKPIPNPLLGLDST). | |
0.1 mL | |
Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5. | |
IgG κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction