Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Epitope Tag Rabbit anti-Virus, Alexa Fluor™ 594, Clone: SV5-P-K, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252653AF594

Catalog No. NP2263A94


Add to cart

Description

Description

V5 Epitope Tag Monoclonal antibody specifically detects V5 Epitope Tag in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence.
Specifications

Specifications

V5 Epitope Tag
SV5-P-K
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence
Conjugated, Purified
Rabbit
IgG Kappa
0.1 mL
Primary
Recognises a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxoVirus simian virus 5.
ELISA, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
V5 epitope tag
Pk (V5) Epitope Tag (GKPIPNPLLGLDST).
Protein A or G purified
Store at 4°C in the dark.
Monoclonal
Virus
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit