Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 tag, anti-Viruses, Clone: SV5-P-K, Abnova™

Monoclonal Antibody

Manufacturer:  Abnova Corporation RAB00098

Catalog No. 89-032-441


Add to cart

Description

Description

Sequence: GKPIPNPLLGLDST
Specifications

Specifications

V5 tag
SV5-P-K
ELISA (0.1 ug/mL) Immunofluorescence Immunohistochemistry (Frozen sections) (5 ug/mL) Immunoprecipitation (10 ug/mL) Radioimmunoassay Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IgG, kappa
RUO
Antibody
Monoclonal
Virus
ELISA, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Radioimmune Assays, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Original antibody is raised against Pk (V5) Epitope Tag.
200 μg
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
This antibody recognizes a 14 amino acid sequence (GKPIPNPLLGLDST) derived from the P and V proteins of the paramyxovirus simian virus 5.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit