Learn More
V5 Tag Monoclonal Antibody (4C12E11), THE™, GenScript™
Mouse Monoclonal Antibody
Supplier: Genscript Corporation A01724200

Description
Reconstitute the lyophilized product with deionized water (or equivalent) to make antibody concentration of 0.5 mg/mL. The V5 epitope tag (GKPIPNPLLGLDST) is derived from a small epitope which presents on the P and V proteins of the paramyxovirus of simian virus 5 (SV5). V5 Tag Antibody is a useful tool in detection of recombinant proteins containing the V5 epitope.THE™ V5 Tag Antibody, mAb, Mouse is produced from the hybridoma resulting from fusion of SP2/0-Ag14 myeloma and B-lymphocytes obtained from mouse immunized with V5 tag peptide conjugated to KLH.
V5 Tag Monoclonal antibody specifically detects V5 Tag in Tag samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western BlotSpecifications
V5 Tag | |
Monoclonal | |
Unconjugated | |
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag | |
V5 tag peptide conjugated to KLH | |
200 μg | |
Primary | |
Antibody | |
IgG2a κ |
ELISA, Flow Cytometry, Immunocytochemistry, Immunoprecipitation, Western Blot | |
4C12E11 | |
PBS with 0.02% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
-20°C or -80°C if preferred | |
Lyophilized | |
Tag |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.