Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

V5 Tag Monoclonal Antibody, Alexa Fluor™ 647, Invitrogen™

Mouse Monoclonal Antibody

Supplier:  Invitrogen 451098

Encompass_Preferred

Catalog No. 45-109-8


Only null left
Add to Cart

Description

Description

The Alexa FluorTM dyes provide for extraordinarily bright antibody conjugates. At the time of preparation, the products are certified to be free of unconjugated dyes and are tested in a cytological experiment to ensure low nonspecific staining. For Alexa FluorTM 647 the approximate fluorescence excitation (Ex) and emission (Em) maxima, in nm: Ex 650/Em 668.

V5 Tag Monoclonal antibody specifically detects V5 Tag in Tag samples. It is validated for Inhibition Assays.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

V5 Tag
Monoclonal
Alexa Fluor 647
simian virus 5 antibody; SPV5gp2; sv5; V5; v-5; V5 Epitope Tag
sequence -Gly-Lys-Pro-Ile-Pro-Asn-Pro-Leu-Leu-Gly-Leu-Asp-Ser-Thr-
80 μL
Primary
Antibody
IgG2a
Inhibition Assays
0.5 mg/mL
PBS with 2mM sodium azide; pH 7.5
Mouse
purified
RUO
4° C, store in dark, DO NOT FREEZE!
Liquid
Tag
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.