Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VABP Antibody (6C239.4), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP224876AF532
Description
VABP Monoclonal specifically detects VABP in Human samples. It is validated for Western Blot.Specifications
VABP | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
KCNIP1 | |
Full length recombinant protein was used as the immunogen. | |
Primary | |
Human | |
IgG1 κ |
Western Blot | |
6C239.4 | |
Western Blot | |
A-type potassium channel modulatory protein 1, KChIP1, KCHIP1MGC95, Kv channel interacting protein 1, Kv channel-interacting protein 1, potassium channel interacting protein 1, Potassium channel-interacting protein 1, VABP, Vesicle APC-binding protein | |
Mouse | |
0.1 mL | |
30820 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction