Learn More
Vaccinia virus Rabbit anti-Virus, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29823
Catalog No. 89-033-668
Add to cart
Description
Description
Rabbit polyclonal antibody raised against vaccinia virus.
  • High specificity
  • High reactivity
Specifications
Specifications
Vaccinia virus
Unconjugated
ELISA Immunofluorescence Western Blot The optimal working dilution should be determined by the end user.
Liquid
Vaccinia virus strain Lister.
500 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Virus
ELISA, Immunofluorescence, Western Blot
Rabbit polyclonal antibody raised against vaccinia virus.
In 0.01 M Phospate buffered saline, pH 7.2 (0.09% sodium azide).
Rabbit
IgG
RUO
Primary
This antibody reacts with all antigens of vaccinia virus. No cross-reactivity with parainfluenza virus type 1, 2, and 3, RSV, adenovirus, influenza A and B virus, or HSV.
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)