Learn More
VAChT/SLC18A3 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: S6-38, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259378FR
Catalog No. NBP259378FR
Add to cart
Description
Description
VAChT/SLC18A3 Monoclonal antibody specifically detects VAChT/SLC18A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
VAChT/SLC18A3
S6-38
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
SLC18A3
Synthetic peptide amino acids 521-532 of human VAChT
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 56kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
MGC12716, solute carrier family 18 (vesicular acetylcholine), member 3, Solute carrier family 18 member 3, solute carrier family 18, member 3, VAChT, VACHTvesicular acetylcholine transporter
Mouse
IgG1
100 ul
Primary
6572
Human, Mouse, Rat