Whatman™ Vacu-Guard Filter: PROMO OFFER

PTFE membrane retains 99.9% of airborn particles ≥0.1µm

Manufacturer: Whatman™ 6722-5001PM

 View all options for this product

Catalog No. 09-744-76PM

  Check Availability
Add to Cart


    Material (Membrane) PTFE
    For Use With (Application) Isolate infectious materials
    Filtration Area (Metric) 16cm2
    Inlet Style 3/8-1/2 in. stepped hose barb
    Outlet Style 3/8-1/2 in. stepped hose barb

    • Installed inline
    • Prevent fluid and aerosol contamination of vacuum pumps or aspiration suction systems
    • Eliminate hazardous exhaust
    • Disposable
    • Filter area: for 50mm diameter

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit