Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vacu-vials Spectrophotometer Reagent Test Kits
$43.25
Specifications
Certifications/Compliance | US EPA, ASTM™, APHA standard procedures |
---|---|
For Use With (Equipment) | Spectrophotometer |
Packaging | 30 Tests/Pack |
Type | Vacu-vials Kit |
Catalog Number | Mfr. No. | Analytes | Includes | Range | Method | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Analytes | Includes | Range | Method | Price | Quantity & Availability | ||
S73104
![]() |
Chemetrics Inc![]() K3503/EMD |
Copper | 30 Ampoules, 25mL sample cup, ampoule blank and instructions | 0 to 7 ppm | Bathocuproine |
Pack of 30 for $43.25
|
|
Description
Easy-to-use chemical test kits work in conjunction with your spectrophotometer to measure the concentration of various chemical species. Absorbance valve can be converted to concentration units with the supplied calibration equation. Each kit contains 30 tests.
- Test methods are based on US EPA, ASTM, and APHA standard procedures for water testing
- Kits have long shelf life (more than two years for most) and pose no storage hazard issues
- All chemicals are self-contained in ampule vials for safety
Specifications
US EPA, ASTM™, APHA standard procedures | |
Spectrophotometer | |
30 Tests/Pack | |
Vacu-vials Kit |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title