Learn More
CHEMetrics Vacu-vials Test Kits
Use with any spectrophotometer that accepts a 13mm cell
$49.25 - $96.00
Catalog Number | Mfr. No. | Analytes | Range | Method | No. of Tests | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Analytes | Range | Method | No. of Tests | Price | Quantity & Availability | ||
13-299-339
|
CHEMetrics
K9923 |
Zinc 2 | 0.6 to 6ppm | Zincon | 30.0 |
Each for $53.25
|
|
13-299-527
|
CHEMetrics
K7003 |
Nitrite | 0.08 to 0.8ppm | Azo dye formation | 30.0 |
Each for $53.25
|
|
13-299-525
|
CHEMetrics
K2513 |
Free and Total Chlorine 2 | 0.4 to 5ppm | DPD (free and total) | 30.0 |
Each for $53.25
|
|
13-299-526
|
CHEMetrics
K6923 |
Nitrate 2 | 0.4 to 3ppm | Cadmium Reduction | 30.0 |
Each for $96.00
|
|
13-299-336
|
CHEMetrics
K9503 |
Sulfide 1 | 0.2 to 3ppm | Methylene Blue | 30.0 |
Each for $53.25
|
|
13-299-302
|
CHEMetrics
K1503 |
Ammonia 1 | 0.5 to 7ppm | Direct nesslerization | 30.0 |
Each for $54.75
|
|
13-299-307
|
CHEMetrics
K2803 |
Chromate 1 | 0 to 3.5ppm | Diphenylcarbazide | 30.0 |
Each for $49.25
|
|
13-299-311
|
CHEMetrics
K3803 |
Cyanide (free) | 0.04 to 0.4ppm | Isonicotinic-Barbituric Acid | 29.0 |
Each for $51.00
|
|
13-299-330
|
CHEMetrics
K8003 |
Phenol 1 | 0.4 to 8ppm | 4-Aminoantipyrine | 30.0 |
Each for $53.25
|
|
13-299-321
|
CHEMetrics
K6023 |
Total Iron 2 | 0 to 2.5ppm | Total PDTS | 30.0 |
Each for $53.25
|
|
13-299-332
|
CHEMetrics
K8503 |
Phosphate 1 | 5 to 80ppm | Vanadomolybdophosphoric Acid | 30.0 |
Each for $53.25
|
|
13-299-334
|
CHEMetrics
K9003 |
Silica | 0.5 to 10ppm | Heteropoly Blue | 30.0 |
Each for $53.25
|
|
13-299-337
|
CHEMetrics
K9523 |
Sulfide 2 | 0.6 to 6ppm | Methylene Blue | 30.0 |
Each for $53.25
|
|
13-299-338
|
CHEMetrics
K9903 |
Zinc 1 | 0.3 to 3ppm | Zincon | 30.0 |
Each for $53.25
|
|
13-299-528
|
CHEMetrics
K7513 |
Dissolved Oxygen 2 | 2 to 15ppm | Indigo Carmine | 30.0 |
Each for $54.75
|
|
13-299-538
|
CHEMetrics
K 8513 |
Phosphate 2 | 0.3 to 8ppm | Stannous Chloride | 30.0 |
Each for $53.25
|
|
13-299-524
|
CHEMetrics
K1523 |
Ammonia 2 | 1 to 14ppm | Direct nesslerization | 30.0 |
Each for $54.75
|
|
13-299-322
|
CHEMetrics
K6903 |
Nitrate 1 | 0.2 to 1.5ppm | Cadmium Reduction | 30.0 |
Each for $96.00
|
|
13-299-324
|
CHEMetrics
K6933 |
Nitrate 3 | 5 to 50ppm | Cadmium Reduction | 30.0 |
Each for $96.00
|
|
13-299-319
|
CHEMetrics
K6003 |
Total and Soluble Iron 1 | 0.2 to 6ppm | Total and Soluble Phenanthroline | 30.0 |
Each for $51.00
|
|
13-299-327
|
CHEMetrics
K7503 |
Dissolved Oxygen 1 | 0.2 to 2ppm | Indigo Carmine | 30.0 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
13-299-320
|
CHEMetrics
K6013 |
Total and Soluble Iron 3 | 1 to 25ppm | Total and Soluble Phenanthroline | 30.0 |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
Description
Snap-and-read Vacu-vials self-filling ampules are the fastest, easiest reagent systems available for precision colorimetric determination of water/wastewater.
- Each vacuum-sealed ampule contains a unit dose of premeasured reagent
- Simply immerse the ampule in the water sample, snap its tip, and an analytically correct amount of sample is pulled inside and mixed
- Sample is now ready to be analyzed by your spectrophotometer (instrument must accept a 13mm cell directly or with an adapter)
- Direct readouts in concentration may be obtained by using CHEMetrics V-2000 Photometer or by preprogramming your spectrophotometer using standard solutions or the data pairs provided in the Vacu-vials instructions
- For immediate results in your spectrophotometer without programming, use supplied equation or calibration tables to convert from absorbance or transmittance to concentration units
Order Info
Complete test kits contain 30 ampules, calibration tables, zero standard, accessory solutions (when necessary), snap cup, and instructions.
Specifications
Vacu-vials™ Test Kit |
Safety and Handling
ShelfLife : 2 Months
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.