Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vacu-vials™ Test Kits

Use with any spectrophotometer that accepts a 13mm cell

Manufacturer:  CHEMETRICS INC K4203

 View more versions of this product

Catalog No. 13-299-313


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Formaldehyde
0.4 to 8ppm
Cardboard box, 30 ampoules, activator solution, 25mL sample cup, ampoule blank, instructions
Purpald
Vacu-vials™ Test Kit
V-2000 photometer or spectrophotometer
30.0

Safety and Handling

ShelfLife : 5 Months

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit