Learn More
BUCHI Vacuum Controller V-850
SDP
Semi-Automated Vacuum Controller V-850 with a LCD-display to control the vacuum by a valve, or the new speed control technology with the Vacuum Pump V-700/V-710.
Manufacturer:  BUCHI 047293
 View more versions of this product
Catalog No. 05-405-038
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Complete and ready to use with an aeration valve and pressure sensor included
  • Timer and laboratory vacuum control (LabVac) functions
  • Reduces water consumption by using a cooling water valve
  • Includes the communication cable for the Rotavapor™
  • Features a solvent library with the 43 most commonly used solvents
Specifications
Specifications
Rotavapor™ R-250
Semi-automatic
Vacuum Controller
V-850