Learn More
BUCHI Vacuum Controller V-850
Semi-Automated Vacuum Controller V-850 with a LCD-display to control the vacuum by a valve, or the new speed control technology with the Vacuum Pump V-700/V-710.
$3940.00 - $4310.00
Specifications
Model V-850
Type Vacuum Controller
Controller Type Semi-automatic
View More Specs
Products 2
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
05-405-039
View Documents
BUCHI
047295
Rotavapor™ R-220
Each for $4,310.00
Add to cart
05-405-038
View Documents
BUCHI
047293
Rotavapor™ R-250
Each for $3,940.00
Add to cart
Description
Description
  • Complete and ready to use with an aeration valve and pressure sensor included
  • Timer and laboratory vacuum control (LabVac) functions
  • Reduces water consumption by using a cooling water valve
  • Includes the communication cable for the Rotavapor™
  • Features a solvent library with the 43 most commonly used solvents
Specifications
Specifications
V-850
Vacuum Controller
Semi-automatic