Learn More
Heidolph™ Vacuum seal
The highly durable and long lasting PTFE vacuum seals with FKM sealing ring achieve maximum tightness and save on long-term spare parts costs.
Manufacturer:  Heidolph™ 036210032
Catalog No. 02-321-080
Add to cart
Description
Description
  • PTFE spare seal with FKM sealing ring
  • The combination of both materials allows for very good chemical resistance, while at the same time offering the best sealing properties
  • Long running time without abrasion
  • Highest density
  • FDA compliant
Specifications
Specifications
PTFE
Evaporators
Vacuum Seal
Natural
FKM sealing ring