Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Pederson Vaginal Speculum, Sklar, Blunt, Angled, Smooth, Medium, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21348
Econo Pederson Vaginal Speculum, Sklar, Blunt, Angled, Smooth, Medium, Stainless Steel. The Econo Pederson Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, pap smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The blades of the Pederson Speculum are narrower than the blades of the Graves Speculum. The practitioner can best determine which type and size speculum is most appropriate for the patient. Econo instruments are ready-to-use, disposable, floor-grade instruments sold in bulk quantities.
Catalog No. 50-143-7614