Learn More
VAI 1700 Mitcool Coveralls
Manufacturer:  VAI 1700ES1728
 View more versions of this product
Catalog No. 19-546-393
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
Elastic wrist and ankle, single stitch sewn thumb loops with bound edges
Elastic
Elastic,Thumb loops
White
Use in Grade A (ISO 5/Class 100) areas or as a personal protection garment
Polypropylene
Yes
Piped,Stitched
Large
Yes