VAI 1700 Mitcool Coveralls

Manufacturer: VAI 1700-E-S-1729

 View more versions of this product

Catalog No. 19-546-394

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Material Polypropylene
    Color White
    For Use With (Application) Use in Grade A (ISO 5/Class 100) areas or as a personal protection garment
    Sterile Yes
    Includes Elastic wrist and ankle, single stitch sewn thumb loops with bound edges
    Seam Type Piped with Reinforced Stitching
    Size X-Large
    Disposable Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit