Learn More
VAI DECON-CYCLE™ II Germicidal Detergent
Phosphate-free
Manufacturer:  VAI DCY032Z
 View more versions of this product
Catalog No. 19-546-815
Add to cart
Includes: Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
Description
Description
A low pH, dilutable, hospital detergent effective in 10 minutes at 20°C in hard water up to 400ppm (calculated as CaCO3) in the presence of 5% blood serum. Recommended for use in pharmaceutical, biotechnology, medical device manufacturing, and in healthcare institutions to control cross-contamination hazards.
  • Multi-phenolic formula designed to clean, disinfect, and deodorize any washable inanimate surface
  • Filtered at 0.2µm and gamma irradiated
  • Strong enough to kill a broad spectrum of pathogenic bacteria, yet mild enough to have no harmful effect on disinfected surfaces
  • Completely traceable and validated for sterility and shelf life
Order Info
Double-bag packaging.
Compliance
Completely tested according to current USP compendium.
Specifications
Specifications
Safety
Kills broad spectrum of pathogenic Bacteria, pharmaceutical, biotechnology, medical device and health care industries
2 oz.
Sterile