VAI Decon-Phase™ Germicidal Solution
Kill a broad spectrum of pathogenic and non-pathogenic bacteria with this potassium-based phenolic germicidal solution
Manufacturer: VAI DPH-03-2Z
 View more versions of this product
Catalog No. 19-546-849
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Certifications/Compliance USP, ISO
    Dilution Non-dilluted
    Filtered 0.2μm
    Includes Certificate of irradiation, certificate of analysis and certificate of sterility
    Size 2 oz.
    Sterile Yes
    For Use With (Application) Used in cleaning rotation cycles where hard surface disinfectants and cleaners are recommended for maintaining a clean environment and controlling the hazards of cross contamination
  • Description & Specifications
Related Products
Product Content Feedback