VAI Decon-Phase™ Germicidal Solution
Kill a broad spectrum of pathogenic and non-pathogenic bacteria with this potassium-based phenolic germicidal solution
Manufacturer: VAI DPH-04-1/2Z
 View all options for this product
Catalog No. 19-547-048
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Size 1 gal.
    Certifications/Compliance USP, ISO
    For Use With (Application) Used in cleaning rotation cycles where hard surface disinfectants and cleaners are recommended for maintaining a clean environment and controlling the hazards of cross contamination
    Dilution 1:256 dillution
    Filtered 0.2μm
    Includes Certificate of irradiation, certificate of analysis and certificate of sterility
    Sterile Yes
    Mild enough to have no harmful effect on the surfaces being disinfected
    • Phosphate-free germicidal detergent
    • Used where a broad spectrum of disinfection is required in moderate amounts of organic soil
    • Completely tested for sterility according to current USP compendium
    • Available in the quadruple bagged ABCD Clean Room Introduction System
    • Filtered at 0.2 microns
    • Double-bagged packaged
    • Gamma irradiated
    Completely traceable
  • Description & Specifications
Related Products
Product Content Feedback