Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VAI DECON-PHENE™ Germicidal Detergent
Sodium-based, phosphate-free phenolic synthetic germicidal detergent
Supplier: VAI DP032Z
Description
Designed for use where a broad-spectrum disinfectant is required in moderate amounts of organic soil such as decontamination of walls, ceilings and floors.
- Filtered at 0.2μm and gamma irradiated to a 10-6 Sterility Assurance Level
- Strong enough to kill a broad spectrum of pathogenic and non-pathogenic bacteria; mild enough to have no harmful effect on disinfected surfaces
- Detergent that has efficacy performance that confirms to AOAC protocol testing at a use dilution of 1:128 in he presence of 5% blood serum at 20°C with an exposure time of 10 minutes
- Completely traceable and validated for sterility and shelf life
Order Info
Double-bagged packaged
Certifications
Lot-specific Certificate of Irradiation, Certificate of Analysis, LAL testing analysis and sterility report
Compliance
Completely tested according to current USP compendium
Specifications
Concentrate | |
ISO 5 | |
2 oz. |
Sterile | |
For use by pharmaceutical and biotechnology industries, medical device manufacturers, hospitals and any healthcare institutions |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction