VAI Easy2Gown™ Mitcool 1700 Cleanroom Coveralls
Proprietary fold reduces the chance of the individual contaminating the outside of the garment
Manufacturer: VAI 1700P-EHB-S-17012
 View more versions of this product
Catalog No. 19-546-415
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Material Polypropylene
    Includes Elastic wrist and ankle, attached hood and boots, doubled stitch sewn thumb loops with bound edges
    Seam Type Piped with Reinforced Stitching
    Color White
    Size 2X-Large
    Disposable Yes
    For Use With (Application) Use in Class 100 (Grade A, ISO 5), Class 10,000 (Grade B, ISO 7) and in Class 100,000 (Grade C, ISO 8) areas in both aseptic and non-aseptic manufacturing and testing operations
    Sterile Yes
    • Bonded polypropylene with laminate
    • Sewn thumb loops to eliminate shifting
    • Piped seams with reinforced stitches
    • Excellent chemical resistance
    • Disposable
  • Description & Specifications
Related Products
Product Content Feedback