VAI Mitcool 1700 Cleanroom Boot Covers
Manufacturer: VAI 1700-MC-NS-S-1790
 View more versions of this product
Catalog No. 19-546-409
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Sole Type Non-skid
    Product Type Boot Covers
    Material Polypropylene
    Seam Type Bound
    Style Mid-calf
    Sterile Yes
    Size Large, X-Large
    For Use With (Application) Ideal for cleanroom use
  • Description & Specifications
Related Products
Product Content Feedback