Learn More
VAI PF-2 Face Masks
Comfortable material masks for use in pharmaceutical, biotechnology, and medical device industries
Manufacturer:  VAI PF-2F-T-4-S-2014
 View more versions of this product
Catalog No. 19-546-445
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
Four ties
Large
Yes
Rayon, Latex Acrylic
99%
0.45μ