VAI PF-2 Face Masks
Comfortable material masks for use in pharmaceutical, biotechnology, and medical device industries
Manufacturer: VAI PF2SMT4S2020
 View more versions of this product
Catalog No. 19-546-451
  Check Availability
Add to Cart
    Specifications
    Includes Four ties
    Material Rayon, Latex Acrylic
    Size One
    Filter Efficiency 99%
    Sterile No
    Pore Size 0.45μ
    • Made from rayon laminated with an acrylic binder for low particulate and shredding characteristics
    • 99% filtration measured at .045 microns
    • Eliminates fogging of goggles
    • Provides excellent breathability
  • Description & Specifications
Related Products
Product Content Feedback