VAI STERI-BUFFER™ Phosphate Buffer

For disinfectant preparation and dilution

Manufacturer: VAI SB10090

 View more versions of this product

Catalog No. 19-546-920

  Check Availability
Add to Cart

    Specifications

    Includes Certificate of analysis, certificate of sterility
    Product Name Phosphate Buffer
    Quantity 72 x 90mL
    Shelf Life 2 Years
    Sterile Yes
    For Use With (Application) For disinfectant preparation and dilution
    pH 7.2

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit