Learn More
Valence Shell Electron Pair Repulsion Models
$10.00
Specifications
For Use With (Application) | Illustrates molecular geometry, resonance and molecule polarity |
---|---|
Type | Valence shell electron pair repulsion model |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
S65371
![]() |
Ryler Enterprises
VSP2 |
Red 4-peg Oxygen atom centers, green 4-peg Halogen atom centers, purple 5-peg Phosphorous atom centers, Silver 6-peg sulfur (xenon) atom centers, 2 yellow 4-peg sulfur atom centers, 1.25 in. PVC, clear bonds (single bonds), 1.625 in. PVC, clear bonds (double bonds), background information booklet, lab procedure, student response form and answer key |
Each for $10.00
|
|
Description
Valence Shell Electron Pair Repulsion (VSEPR) models illustrate molecular geometry, resonance, and molecule polarity.
VSEPR 1 model supports students' investigation of the octet rule to create models of molecules with linear, tetrahedral, pyramidal, bent, and trigonal planar shapes.Model includes:
- Two carbons
- Four oxygens
- Five hydrogens
- Three halogens
- Three nitrogens
- Two sulfurs
- Six short tubes
- Six long tubes
Model includes:
- 2-2 trigonal bipyramidal
- Two octahedral
- Five oxygens
- Three hydrogens
- Seven halogens
- Two nitrogens
- Eight short tubes
- Eight long tubes
Specifications
Illustrates molecular geometry, resonance and molecule polarity | |
Valence shell electron pair repulsion model |