Valeo™ Washable Half-Finger Gloves
Reduce shock and stress from vibration
Manufacturer: Valeo Inc GLASLHMED
 View all options for this product
Catalog No. 19-048-681B
Please call customer service for current pricing. 1-800-766-7000 
    Material Leather
    Size Medium
    • AVG™ vibration-absorbing padding in palm, thumb, and index finger areas
    • Leather glove has nylon mesh back
    • Available with elastic wrist wrap with a nonconductive support pad or adjustable hook and loop closure for a nonrestrictive fit
    • Washable
    • Black
  • Description & Specifications
Related Products
Product Content Feedback