Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Valeric acid, ≥99.8% (GC), MilliporeSigma™ Supelco™
$65.75 - $114.00
Specifications
CAS | 109-52-4 |
---|---|
Type | Analytical Standard |
Linear Formula | CH3(CH2)3 COOH |
MDL Number | MFCD00004413 |
Synonym | n-Valeric acid; Pentanoic acid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
11-102-0222
|
MilliporeSigma Supelco
750541ML |
1 mL |
Each for $65.75
|
|
|||||
11-101-9492
|
MilliporeSigma Supelco
750545ML |
5 mL |
Each for $114.00
|
|
|||||
Description
Cleaning Products, Cosmetics, Food and Beverages and Personal CareSpecifications
109-52-4 | |
CH3(CH2)3 COOH | |
n-Valeric acid; Pentanoic acid | |
≥99.8% (GC) | |
707°F | |
3.5 (vs. air) | |
-20°C to -18°C | |
96°C |
Analytical Standard | |
MFCD00004413 | |
102.13 | |
Analytical Standard | |
0.15 mmHg (20°C) | |
0.939 g/mL (at 25°C) | |
110°C to 111°C (10 mmHg); 185°C | |
Neat |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : YV6100000
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title