Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

γ-Valerolactone, 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
100mL
500mL
Packaging:
Glass bottle
108-29-2
C5H8O2
100.12
MFCD00005400
GAEKPEKOJKCEMS-UHFFFAOYSA-N
gamma-valerolactone,4-valerolactone,4-pentanolide,5-methyldihydrofuran-2 3h-one,4-hydroxypentanoic acid lactone,gamma-pentalactone,4-hydroxyvaleric acid lactone,2 3h-furanone, dihydro-5-methyl,4-methyl-gamma-butyrolactone,pentanolide-1,4
7921
CHEBI:48569
5-methyloxolan-2-one
CC1CCC(=O)O1
108-29-2
C5H8O2
100.12
MFCD00005400
GAEKPEKOJKCEMS-UHFFFAOYSA-N
gamma-valerolactone,4-valerolactone,4-pentanolide,5-methyldihydrofuran-2 3h-one,4-hydroxypentanoic acid lactone,gamma-pentalactone,4-hydroxyvaleric acid lactone,2 3h-furanone, dihydro-5-methyl,4-methyl-gamma-butyrolactone,pentanolide-1,4
7921
CHEBI:48569
5-methyloxolan-2-one
CC1CCC(=O)O1
Viscosity 2.18 mPa.s (25°C)
Quantity 5mL
Specific Gravity 1.05
Packaging Glass bottle
Flash Point 96°C
Infrared Spectrum Authentic
Refractive Index 1.4320 to 1.4340
Melting Point -31.0°C
Density 1.0500g/mL
Boiling Point 207.0°C to 208.0°C
Color Colorless
Assay Percent Range 98%
Beilstein 17, 235
Solubility Information Solubility in water: miscible. Other solubilities: miscible with most common organic solvents,resins,propylene glycol,oil
Formula Weight 100.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material γ-Valerolactone
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.