Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valerophenone 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
1009-14-9
C11H14O
162.23
MFCD00009480
XKGLSKVNOSHTAD-UHFFFAOYSA-N
valerophenone,butyl phenyl ketone,pentanophenone,1-phenyl-1-pentanone,1-pentanone, 1-phenyl,unii-f27q043nt1,n-butyl phenyl ketone,1-benzoylbutane,valerophenone, 8ci
66093
CHEBI:36812
1-phenylpentan-1-one
CCCCC(=O)C1=CC=CC=C1
1009-14-9
C11H14O
162.23
MFCD00009480
XKGLSKVNOSHTAD-UHFFFAOYSA-N
valerophenone,butyl phenyl ketone,pentanophenone,1-phenyl-1-pentanone,1-pentanone, 1-phenyl,unii-f27q043nt1,n-butyl phenyl ketone,1-benzoylbutane,valerophenone, 8ci
66093
CHEBI:36812
1-phenylpentan-1-one
CCCCC(=O)C1=CC=CC=C1
Infrared Spectrum Authentic
Melting Point -9.0°C
Flash Point 102°C
Refractive Index 1.5143 to 1.5163
Packaging Glass bottle
Density 0.9800g/mL
Specific Gravity 0.98
Boiling Point 105.0°C to 107.0°C (5.0 mmHg)
Quantity 100g
Color Green-Yellow to Yellow
Assay Percent Range 98%
Linear Formula C6H5CO(CH2)3CH3
Beilstein 07,327
Formula Weight 162.23
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Valerophenone, 98%
Show More Show Less

EINECSNumber : 213-767-3

TSCA : TSCA