Alfa Aesar™ Valerophenone p-toluenesulfonylhydrazone, 97%

Manufacturer: Alfa Aesar™ H5339214

Quantity 25g

 View more versions of this product

Catalog No. AAH5339214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 69015-74-3
    Molecular Formula C18H22N2O2S
    Formula Weight 330.44
    MDL Number MFCD00159338
    Synonym Valerophenone tosylhydrazone; p-Toluenesulfonic acid (1-phenylpentylidene)hydrazide
    Chemical Name or Material Valerophenone p-toluenesulfonylhydrazone

    Specifications

    Formula Weight 330.44
    Molecular Formula C18H22N2O2S
    MDL Number MFCD00159338
    Synonym Valerophenone tosylhydrazone; p-Toluenesulfonic acid (1-phenylpentylidene)hydrazide
    Melting Point 128° to 136°C
    Quantity 25g
    CAS 69015-74-3
    Chemical Name or Material Valerophenone p-toluenesulfonylhydrazone
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit