Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Valeryl chloride, 98%

Manufacturer:  Alfa Aesar™ B2263718

Quantity 50g

 View more versions of this product

Catalog No. AAB2263718

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
Valeryl chloride is an intermediate for pharmaceuticals, agrochemicals, surfactants, fragrances, electronic chemicals.

Solubility
Hydrolyzes in water. Soluble in ether, chloroform.

Notes
Moisture sensitive. Stay away from oxidizing agents, bases, alcohols.
Description & Specifications

Chemical Identifiers

638-29-9
120.58
UN2502
Pentanoyl chloride
CH3(CH2)3COCl
MFCD00000757
506293
Valeryl chloride

Specifications

638-29-9
120.58
UN2502
Pentanoyl chloride
125° to 127°C
32°C (89°F)
Valeryl chloride
CH3(CH2)3COCl
MFCD00000757
506293
−110°C
50g
Acrid
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318-H226-H302-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: VALERYL CHLORIDE

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit