Alfa Aesar™ Valeryl chloride, 98%

Manufacturer: Alfa Aesar™ B2263730

Quantity 250g

 View more versions of this product

Catalog No. AAB2263730

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 638-29-9
    Molecular Formula CH3(CH2)3COCl
    Formula Weight 120.58
    MDL Number MFCD00000757
    UN Number UN2502
    Beilstein 506293
    Synonym Pentanoyl chloride
    Chemical Name or Material Valeryl chloride

    Specifications

    Formula Weight 120.58
    TSCA TSCA
    Molecular Formula CH3(CH2)3COCl
    MDL Number MFCD00000757
    CAS 638-29-9
    Health Hazard 1 Danger
    Boiling Point 125° to 127°C
    Flash Point 32°C (89°F)
    Odor Acrid
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: VALERYL CHLORIDE
    UN Number UN2502
    Beilstein 506293
    Synonym Pentanoyl chloride
    Melting Point −110°C
    Quantity 250g
    Health Hazard 2 GHS H Statement
    H314-H318-H226-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material Valeryl chloride
    Assay 98%

    Description

    Applications
    Valeryl chloride is an intermediate for pharmaceuticals, agrochemicals, surfactants, fragrances, electronic chemicals.

    Solubility
    Hydrolyzes in water. Soluble in ether, chloroform.

    Notes
    Moisture sensitive. Stay away from oxidizing agents, bases, alcohols.

    GHS H Statement
    H314-H318-H226-H302-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flammable liquid and vapour.
    Harmful if swallowed.
    Harmful if inhaled.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit