Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Validamycin A, 100mg/mL in DMSO, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
37248-47-8
C20H35NO13
497.494
MFCD09028089
JARYYMUOCXVXNK-CSLFJTBJSA-N
443629
CHEBI:29703
(2R,3R,4S,5S,6R)-2-[(1R,2R,3S,4S,6R)-2,3-dihydroxy-6-(hydroxymethyl)-4-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]cyclohexyl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C1C(C(C(C(C1NC2C=C(C(C(C2O)O)O)CO)O)O)OC3C(C(C(C(O3)CO)O)O)O)CO
37248-47-8
C20H35NO13
497.494
MFCD09028089
JARYYMUOCXVXNK-CSLFJTBJSA-N
443629
CHEBI:29703
(2R,3R,4S,5S,6R)-2-[(1R,2R,3S,4S,6R)-2,3-dihydroxy-6-(hydroxymethyl)-4-[[(1S,4R,5S,6S)-4,5,6-trihydroxy-3-(hydroxymethyl)cyclohex-2-en-1-yl]amino]cyclohexyl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C1C(C(C(C(C1NC2C=C(C(C(C2O)O)O)CO)O)O)OC3C(C(C(C(O3)CO)O)O)O)CO
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Validamycin A, 100mg/mL in DMSO, sterile-filtered

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : -30° to -10°C