Learn More
Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Manufacturer:  Thermo Scientific™ 840257300
 View more versions of this product
Catalog No. 84-025-7300
Description
Description
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Solutions include:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance where applicable
  • Installation and instrument certification
Solutions available for GENESYS 40-180, BioMate 160, Evolution200 series, Evolution 350, NanoDrop One, NanoDrop 2000, and NanoDrop 8000 instruments
Specifications
Specifications
Qualification Filter Kit for NanoDrop OneC and NanoDrop 2000C Spectrophotometers
NanoDrop OneC and NanoDrop 2000C Spectrophotometers
Includes: 1 x Filter holder blank, 1 x 10%T Filter, Calibrated, 1 x 50%T Filter, Calibrated, Certificate with traceability and NVLAP 17025 certification