Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Valinomycin, 10mg/mL in DMSO, sterile-filtered, Alfa Aesar™

Click to view available options
Quantity:
5 x 1mL
2001-95-8
C54H90N6O18
1111.338
MFCD00005114
FCFNRCROJUBPLU-CVZFCLRKSA-N
131668508
(3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
CC1C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)O1)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C
2001-95-8
C54H90N6O18
1111.338
MFCD00005114
FCFNRCROJUBPLU-CVZFCLRKSA-N
131668508
(3S,6S,12R,15S,18S,21R,24R,27S,30S,33R)-6,18,30-trimethyl-3,9,12,15,21,24,27,33,36-nona(propan-2-yl)-1,7,13,19,25,31-hexaoxa-4,10,16,22,28,34-hexazacyclohexatriacontane-2,5,8,11,14,17,20,23,26,29,32,35-dodecone
CC1C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)OC(C(=O)NC(C(=O)O1)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C)C)C(C)C)C(C)C)C(C)C
Beilstein 78657
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Valinomycin, 10mg/mL in DMSO, sterile-filtered

GHS H Statement
H302-H312-H227
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.

GHS P Statement
P210-P280-P301+P312-P312-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : YV9468000

TSCA : TSCA

storageNote1 : -30° to -10°C