Learn More
Valpha TCR, anti-Human, Clone: 34, Abnova™
Monoclonal Antibody
Manufacturer:  Abnova Corporation RAB00027
Catalog No. 89-032-370
Add to cart
Specifications
Specifications
Valpha TCR
34
ELISA (0.1 ug/mL) Flow Cytometry (1 ug/mL) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
IgG1, kappa
RUO
Antibody
Monoclonal
Human
ELISA, Flow Cytometry, Western Blot
Unconjugated
In PBS, pH 7.2 (0.02% Proclin 300).
Original antibody is raised against purified recombinant soluble 85.33 scTCR.
200 μg
Store at 4°C for up to 3 months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Primary
This antibody binds specifically to the recombinant and native variable domain of the alpha chain which comprises the T-cell receptor.